Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC286178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134276.1 | 2 | 769 | |||
GenBank mRNA | AK090741.1 | - | 2 | 694 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000518934.1 | - | 2 | 769 | Chr. 8 - 57746149 - 57750199 [+] on Build GRCh38 |