Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105006.1 | 6 | 1513 | |||
GenBank mRNA | BC015449.1 | - | 6 | 1513 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000518732.1 | - | 6 | 1513 | Chr. 8 - 76406654 - 76524356 [+] on Build GRCh38 |