Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR100HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024430.2 | 4 | 1922 | |||
NR_137175.1 | 2 | 2193 | ||||
NR_137176.1 | 2 | 1904 | ||||
NR_137177.1 | 2 | 1883 | ||||
NR_137179.1 | 5 | 1876 | ||||
NR_137180.1 | 5 | 1934 | ||||
NR_137181.1 | 3 | 1819 | ||||
NR_137182.1 | 4 | 1901 | ||||
NR_137183.1 | 3 | 1810 | ||||
NR_137184.1 | 2 | 1728 | ||||
NR_137185.1 | 3 | 1789 | ||||
NR_137186.1 | 3 | 1698 | ||||
NR_137187.1 | 3 | 1677 | ||||
NR_137188.1 | 3 | 1767 | ||||
NR_137189.1 | 2 | 2214 | ||||
NR_137190.1 | 3 | 2247 | ||||
NR_137191.1 | 3 | 2226 | ||||
NR_137192.1 | 4 | 2086 | ||||
NR_137193.1 | 5 | 1913 | ||||
NR_137194.1 | 3 | 1788 | ||||
NR_137195.1 | 2 | 1956 | ||||
NR_137196.1 | 2 | 1977 | ||||
GenBank mRNA | AB075498.1 | - | 2 | 2446 | ||
AK021552.1 | - | 1 | 1088 | |||
AK123947.1 | - | 2 | 2130 | |||
AL832469.1 | - | 3 | 1482 | |||
BC040599.1 | - | 2 | 1506 | |||
BX538058.1 | - | 2 | 1484 | |||
BX647608.1 | - | 2 | 1484 |