Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102723330 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132279.1 | 2 | 434 | |||
GenBank mRNA | BX103002.1 | - | 2 | 434 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000529781.1 | - | 2 | 435 | Chr. 11 - 17695267 - 17696940 [-] on Build GRCh38 |