Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120477.1 | 3 | 564 | |||
GenBank mRNA | BC031884.1 | - | 3 | 575 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000536518.1 | - | 3 | 564 | Chr. 12 - 5238048 - 5243151 [-] on Build GRCh38 |