Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110058.1 | 5 | 1557 | |||
GenBank mRNA | BC040912.1 | - | 5 | 1557 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000542578.1 | - | 5 | 1557 | Chr. 12 - 129208601 - 129212662 [+] on Build GRCh38 |