Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038358.1 | 1 | 101 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000556479.5 | - | 1 | 101 | Chr. 14 - 51333393 - 51365557 [+] on Build GRCh38 |