Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720250.3 | XP_006720313.1 | 4 | 3159 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000556640.1 | - | 1 | 223 | Chr. 14 - 64422935 - 64448557 [-] on Build GRCh38 |