Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120363.1 | 3 | 759 | |||
GenBank mRNA | BC041477.1 | - | 3 | 759 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558208.1 | - | 3 | 759 | Chr. 15 - 80554609 - 80562944 [-] on Build GRCh38 |