Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 7 | 753 | |||
NR_110481.1 | 5 | 485 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546201.1 | XP_011544503.1 | 5 | 512 | ||
XM_011546205.1 | XP_011544507.1 | 9 | 923 | |||
XM_011546206.1 | XP_011544508.1 | 8 | 900 | |||
XM_017030115.1 | XP_016885604.1 | 1 | 12 |
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK097598.1 | - | 6 | 902 |