Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00928 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027074.1 | 3 | 1856 | |||
NR_027075.1 | 4 | 974 | ||||
NR_027076.1 | 3 | 687 | ||||
NR_027077.1 | 3 | 858 | ||||
GenBank mRNA | BC039350.1 | - | 3 | 1857 | ||
BC102002.1 | - | 3 | 657 | |||
BC102003.1 | - | 4 | 693 | |||
BC102004.1 | - | 4 | 406 | |||
BC102005.1 | - | 3 | 658 | |||
BC113965.1 | - | 3 | 577 | |||
BX103832.1 | - | 1 | 146 |