Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINGO1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120361.1 | 3 | 405 | |||
NR_120362.1 | 3 | 387 | ||||
GenBank mRNA | BC038758.1 | - | 3 | 403 | ||
BI463387.1 | - | 3 | 408 | |||
HY007517.1 | - | 3 | 376 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000557799.1 | - | 3 | 404 | Chr. 15 - 77660052 - 77668074 [+] on Build GRCh38 |