Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929679 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120366.1 | 4, 5 | 562 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000557908.2 | - | 1, 2 | 424 | Chr. 15 - 85754941 - 85756237 [-] on Build GRCh38 |