Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110372.1 | 5 | 473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558797.1 | - | 5 | 473 | Chr. 15 - 66582190 - 66685794 [+] on Build GRCh38 |