Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100996338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126340.1 | 2, 3 | 495 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000561336.2 | - | 2, 3 | 362 | Chr. 16 - 54245620 - 54270746 [-] on Build GRCh38 |