Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01583 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120367.1 | 2 | 150 | |||
GenBank mRNA | BC043257.2 | - | 2 | 150 | ||
BX104219.1 | - | 2 | 113 |