Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120324.1 | 5 | 970 | |||
GenBank mRNA | BC038379.1 | - | 5 | 970 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000558621.2 | - | 3 | 970 | Chr. 15 - 97370371 - 97427952 [-] on Build GRCh38 |