Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC283710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243538.1 | NP_001230467.1 | 1 | 106 | ||
XM_011521463.2 | XP_011519765.1 | 1 | 105 | |||
XM_011521465.2 | XP_011519767.1 | 1 | 105 | |||
XM_017022084.1 | XP_016877573.1 | 1 | 105 | |||
GenBank mRNA | BF974240.1 | - | 1 | 107 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ641953.1 | - | 83 | Chr. - - [-] on Build |