Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120378.1 | 1, 2 | 18 | |||
GenBank mRNA | DC422276.1 | - | 1, 2 | 18 |
Gene Symbol: LOC103171574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120379.1 | 1, 2 | 117 | |||
GenBank mRNA | AK311032.1 | - | 1, 2 | 18 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000559866.1 | - | 1, 2 | 18 | Chr. 15 - 84422618 - 84425882 [+] on Build GRCh38 |