Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929660 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130777.1 | 7 | 1062 | |||
GenBank mRNA | AK098570.1 | - | 7 | 1079 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000602740.3 | - | 2 | 172 | Chr. 5 - 29164425 - 29167797 [+] on Build GRCh38 |