Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134605.1 | 2 | 453 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000568654.1 | - | 2 | 466 | Chr. 18 - 36179996 - 36187448 [-] on Build GRCh38 |