Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCAT47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131947.1 | 4 | 1771 | |||
NR_131948.1 | 3 | 1657 | ||||
GenBank mRNA | AK093002.1 | - | 3 | 1657 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000562231.5 | - | 3 | 1656 | Chr. 16 - 80828735 - 80892595 [-] on Build GRCh38 |