Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLGAP1-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038895.1 | 3 | 672 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000577649.1 | - | 3 | 672 | Chr. 18 - 3878180 - 3897069 [+] on Build GRCh38 |