Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100128006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104607.1 | 2 | 2755 | |||
GenBank mRNA | BX649083.1 | - | 2 | 2703 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000584772.1 | - | 2 | 2713 | Chr. 17 - 12760140 - 12790242 [-] on Build GRCh38 |