Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134645.1 | 2 | 155 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000577327.1 | - | 2 | 155 | Chr. 18 - 6728821 - 6729862 [-] on Build GRCh38 |