Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCAT19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040109.2 | 4 | 1448 | |||
NR_136334.1 | 3 | 560 | ||||
GenBank mRNA | AK309432.1 | - | 3 | 560 | ||
BC070102.1 | - | 4 | 1444 |