Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01255 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110778.1 | 2, 3 | 307 | |||
NR_110779.1 | 3, 4 | 228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000586947.5 | - | 2, 3 | 245 | Chr. 18 - 11490110 - 11506983 [+] on Build GRCh38 |