Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC105371899 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136416.1 | 2 | 367 | |||
NR_136417.1 | 2 | 367 | ||||
GenBank mRNA | BM700476.1 | - | 2 | 265 | ||
CD173460.1 | - | 2 | 141 | |||
CD675441.1 | - | 2 | 208 | |||
CD675892.1 | - | 2 | 324 | |||
DB157978.1 | - | 2 | 367 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000563583.1 | - | 2 | 265 | Chr. 17 - 76962892 - 76969097 [-] on Build GRCh38 |