Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110687.1 | 3 | 798 | |||
NR_110688.1 | 3 | 754 | ||||
NR_110689.1 | 3 | 713 | ||||
NR_110690.1 | 2 | 375 | ||||
NR_110691.1 | 2 | 331 | ||||
GenBank mRNA | BC024732.1 | - | 2 | 304 | ||
BX426127.2 | - | 3 | 650 | |||
CN415214.1 | - | 3 | 697 | |||
HY135890.1 | - | 2 | 236 |