Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046174.2 | 3 | 305 | |||
NR_046454.1 | 3 | 305 | ||||
NR_046455.1 | 2 | 142 | ||||
NR_046456.1 | 3 | 305 | ||||
NR_046457.1 | 3 | 305 | ||||
NR_046458.1 | 3 | 305 | ||||
GenBank mRNA | AB593072.1 | - | 3 | 305 | ||
AB593073.1 | - | 2 | 142 | |||
AB593074.1 | - | 3 | 303 | |||
AB593075.1 | - | 3 | 303 | |||
AB593076.1 | - | 3 | 303 | |||
BC021928.1 | - | 3 | 270 | |||
BC040725.2 | - | 3 | 284 |