Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP09025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109783.1 | 2 | 2532 | |||
GenBank mRNA | AB593148.1 | - | 3 | 2430 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000592718.1 | - | 2 | 2532 | Chr. 17 - 79707266 - 79712317 [+] on Build GRCh38 |