Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC374890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135233.1 | 2 | 722 | |||
GenBank mRNA | BC030765.2 | - | 2 | 722 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000598042.1 | - | 2 | 722 | Chr. 19 - 22615557 - 22623971 [-] on Build GRCh38 |