Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110732.1 | 3 | 612 | |||
GenBank mRNA | BC045766.1 | - | 3 | 612 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000600074.1 | - | 3 | 612 | Chr. 19 - 51152923 - 51181966 [-] on Build GRCh38 |