Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN16-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103455.1 | 3 | 1982 | |||
NR_103456.1 | 2 | 1785 | ||||
GenBank mRNA | AK127889.1 | - | 3 | 1960 | ||
BC043177.2 | - | 2 | 1335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000602810.1 | - | 2 | 1335 | Chr. 6 - 28121798 - 28136866 [-] on Build GRCh38 |