Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC63930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310606.1 | - | 2 | 492 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000606208.5 | - | 2 | 492 | Chr. 20 - 63009383 - 63084133 [+] on Build GRCh38 |