Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC63930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033370.1 | 3 | 1793 | |||
GenBank mRNA | AK128329.1 | - | 3 | 1792 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000607802.1 | - | 3 | 1793 | Chr. 20 - 63009383 - 63085071 [+] on Build GRCh38 |