Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051977.1 | 1, 2 | 224 | |||
GenBank mRNA | AK098538.1 | - | 1, 2 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000607593.1 | - | 1, 2 | 224 | Chr. 13 - 106617810 - 106631359 [+] on Build GRCh38 |