Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051977.1 | 4 | 1629 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000607593.1 | - | 4 | 1629 | Chr. 13 - 106617810 - 106631359 [+] on Build GRCh38 |