Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110304.1 | 3 | 661 | |||
GenBank mRNA | AK307176.1 | - | 3 | 661 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000609200.1 | - | 3 | 661 | Chr. 10 - 75409157 - 75411842 [+] on Build GRCh38 |