Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101929608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109878.1 | 5 | 1200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624692.1 | - | 1200 | Chr. - - [-] on Build |