Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm5893 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045096.1 | 8 | 1588 | |||
GenBank mRNA | BC100485.1 | - | 8 | 1588 | 30919624 |