Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Meg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_027651.2 | 4 | 984 | |||
NR_027652.1 | 4 | 1028 | ||||
GenBank mRNA | AK145456.1 | - | 3 | 1185 | ||
AK155982.1 | - | 3 | 1186 | |||
AK157178.1 | - | 3 | 1186 | |||
BC024818.1 | - | 2 | 356 | |||
BC094421.1 | - | 4 | 973 | |||
Y13832.1 | - | LIT3453 Gtl2 |
4 | 1019 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI730909.1 | - | 2 | 357 | Chr. 12 - 109544446 - 109558310 [+] on Build GRCm38 |