Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 8430429K09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045275.1 | 1, 2 | 110 | |||
NR_045276.1 | 1, 2 | 269 | ||||
GenBank mRNA | AK018450.1 | - | 1, 2 | 110 | ||
BY069744.1 | - | 1, 2 | 270 | |||
BY284012.1 | - | 1, 2 | 318 | |||
BY331368.1 | - | 1, 2 | 272 |