Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810032G03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015579.1 | 392 | ||||
GenBank mRNA | AK012851.1 | - | 1027 | |||
AK028030.1 | - | 912 | ||||
AK029678.1 | - | 1150 | ||||
BC065777.1 | - | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_028318.1 | - | 1150 | Chr. - - [-] on Build |