Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700046A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037693.1 | 1 | 230 | |||
GenBank mRNA | AK012381.1 | - | 1 | 159 | ||
AK020725.1 | - | 1 | 231 | |||
BC072581.1 | - | 1 | 29 | |||
CJ067024.1 | - | 1 | 199 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ572624.1 | - | 1 | 30 | Chr. 18 - 62753176 - 62756146 [-] on Build GRCm38 |