Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5031434C07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045986.1 | 2 | 361 | |||
NR_045987.1 | 2 | 361 | ||||
GenBank mRNA | AK087115.1 | - | 2 | 335 | ||
AK165118.1 | - | 362 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177012.1 | - | 335 | Chr. - - [-] on Build |