Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4931431C16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045807.1 | 4 | 1453 | |||
GenBank mRNA | AK016496.1 | - | 4 | 1423 | ||
AK030185.1 | - | 4 | 1455 |