Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap33os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036630.1 | 12 | 2204 | |||
GenBank mRNA | BC065044.1 | - | 12 | 2203 | 5685982 |