Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003210.1 | - | 4 | 640 | ||
AK012674.1 | - | 4 | 926 | |||
AK014691.1 | - | 4 | 930 | |||
AK151972.1 | - | 4 | 937 | |||
AK168416.1 | - | 802 | ||||
AK219167.1 | - | 1 | 174 | |||
BC026750.1 | - | 4 | 915 | 5135841 |