Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G530011O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029457.1 | 1619 | ||||
GenBank mRNA | AK134468.1 | - | 94 | |||
AK149694.1 | - | 1618 | ||||
AK149709.1 | - | 1619 | ||||
AK149987.1 | - | 1617 |